Learn More
MSC SPI Pin Gage, Black Oxide Plus, 0.271 in, RC 60-62
Catalog No. 5000510861
Encompass
This pin gage features a black oxide coating for corrosion prevention and wear indication. It is constructed from hardened drill rod with a Rockwell C hardness of 60-62 and has an overall length of 2 inches. This precision tool is designed for accurate measurement and inspection tasks.
  • Black oxide coating prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length: 2 in
Catalog No. 50-005-10861 Supplier MSC Supplier No. 84501105
This pin gage features a black oxide coating for corrosion prevention and wear indication. It is constructed from hardened drill rod with a Rockwell C hardness of 60-62 and has an overall length of 2 inches. This precision tool is designed for accurate measurement and inspection tasks.
  • Black oxide coating prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length: 2 in