Learn More
MSC SPI Pipe Thread Plug Gage, Tapered, 3/8-18, Class L-1, Single End
Catalog No. 5000531980
Encompass
This pipe thread plug gage is designed for inspecting internal taper pipe threads to ensure proper assembly of mating parts. It features a tapered design with a 3/8-18 thread size and Class L-1 classification. Made from steel, this single end gage includes a NIST traceability certificate.
  • Designed for inspecting internal taper pipe threads
  • Ensures assembly of mating parts
  • Features a tapered design
  • Has a 3/8-18 thread size
  • Classified as Class L-1
  • Single end configuration
  • Constructed from steel
  • Includes a NIST traceability certificate
  • Conforms to ANSI B1.20.1 standards
  • Suitable for inspecting standard pipe and pipe fittings
Catalog No. 50-005-31980 Supplier MSC Supplier No. 75889865
This pipe thread plug gage is designed for inspecting internal taper pipe threads to ensure proper assembly of mating parts. It features a tapered design with a 3/8-18 thread size and Class L-1 classification. Made from steel, this single end gage includes a NIST traceability certificate.
  • Designed for inspecting internal taper pipe threads
  • Ensures assembly of mating parts
  • Features a tapered design
  • Has a 3/8-18 thread size
  • Classified as Class L-1
  • Single end configuration
  • Constructed from steel
  • Includes a NIST traceability certificate
  • Conforms to ANSI B1.20.1 standards
  • Suitable for inspecting standard pipe and pipe fittings