Learn More
MSC SPI Plug Gage, 0.0600 in Diameter, 0.000200 in Tolerance, Class ZZ Plus
Catalog No. 5000530530
Encompass
The Class ZZ Plus Plug Gage is a precision measuring tool with a diameter of 0.0600 in and a tolerance of 0.000200 in. Constructed from durable steel, this plug gage is designed for accurate measurement applications.
  • Precision measuring tool for accurate applications
  • Features a 0.0600 in diameter
  • Offers a tolerance of 0.000200 in
  • Constructed from durable steel
  • Class ZZ Plus gage for specific measurement requirements
Catalog No. 50-005-30530 Supplier MSC Supplier No. 78382454
The Class ZZ Plus Plug Gage is a precision measuring tool with a diameter of 0.0600 in and a tolerance of 0.000200 in. Constructed from durable steel, this plug gage is designed for accurate measurement applications.
  • Precision measuring tool for accurate applications
  • Features a 0.0600 in diameter
  • Offers a tolerance of 0.000200 in
  • Constructed from durable steel
  • Class ZZ Plus gage for specific measurement requirements