Learn More
MSC SPI Plug Gage, 0.1320 in, Class ZZ Minus, Steel, 0.0002 in
Catalog No. 5000528875
Encompass
This Class ZZ Minus Plug Gage is precisely engineered for accurate measurement of internal diameters. With a nominal diameter of 0.1320 inches and a tolerance of 0.0002 inches, this steel gage ensures precise fit and function verification. It is designed for inspection of hole sizes, locations, and depths.
  • Precisely engineered for accurate measurement of internal diameters.
  • Nominal diameter of 0.1320 inches.
  • Tolerance of 0.0002 inches.
  • Constructed from durable steel for long-lasting performance.
  • Ensures precise fit and function verification.
  • Ideal for inspection of hole sizes, locations, and depths.
  • Manufactured to Class ZZ Minus standards for precision.
Catalog No. 50-005-28875 Supplier MSC Supplier No. 56244809
This Class ZZ Minus Plug Gage is precisely engineered for accurate measurement of internal diameters. With a nominal diameter of 0.1320 inches and a tolerance of 0.0002 inches, this steel gage ensures precise fit and function verification. It is designed for inspection of hole sizes, locations, and depths.
  • Precisely engineered for accurate measurement of internal diameters.
  • Nominal diameter of 0.1320 inches.
  • Tolerance of 0.0002 inches.
  • Constructed from durable steel for long-lasting performance.
  • Ensures precise fit and function verification.
  • Ideal for inspection of hole sizes, locations, and depths.
  • Manufactured to Class ZZ Minus standards for precision.