Learn More
MSC SPI Plug Gage, 0.252 inch Diameter, 0.0002 inch Tolerance, Black Oxide
Catalog No. 5000527516
Encompass
This Class ZZ Plus Plug Gage features a 0.252 inch diameter and a black oxide coating. Constructed from hardened drill rod, this gage is designed to prevent corrosion and clearly indicate wear, ensuring accurate measurements for various applications.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod RC 60-62.
  • Overall length is 2 inches.
Catalog No. 50-005-27516 Supplier MSC Supplier No. 84500917
This Class ZZ Plus Plug Gage features a 0.252 inch diameter and a black oxide coating. Constructed from hardened drill rod, this gage is designed to prevent corrosion and clearly indicate wear, ensuring accurate measurements for various applications.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod RC 60-62.
  • Overall length is 2 inches.