Learn More
MSC SPI Plug Gage, 0.3080 Dia, 0.000200 Tolerance, Steel
Catalog No. 5000530827
Encompass
This Class ZZ Plus Plug Gage is engineered for precise dimensional inspection. It features a 0.3080 inch diameter and a strict 0.000200 inch tolerance, ensuring high accuracy for critical measurements. Constructed from durable steel, this gage provides reliable performance in demanding environments.
  • Measures hole diameters with high precision.
  • Constructed from durable steel for extended use.
  • Maintains a tight tolerance of 0.000200 "
  • Suitable for various quality control and inspection applications.
Catalog No. 50-005-30827 Supplier MSC Supplier No. 78384930
This Class ZZ Plus Plug Gage is engineered for precise dimensional inspection. It features a 0.3080 inch diameter and a strict 0.000200 inch tolerance, ensuring high accuracy for critical measurements. Constructed from durable steel, this gage provides reliable performance in demanding environments.
  • Measures hole diameters with high precision.
  • Constructed from durable steel for extended use.
  • Maintains a tight tolerance of 0.000200 "
  • Suitable for various quality control and inspection applications.