Learn More
MSC SPI Plug Gage, 0.3140 in, Class ZZ Plus, Black Oxide
Catalog No. 5000527517
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement with a diameter of 0.3140 inches. It features a durable black oxide coating, providing reliable performance for various applications.
  • Precise measurement with a diameter of 0.3140 inch
  • Class ZZ Plus tolerance for accuracy
  • Durable black oxide coating for enhanced longevity
Catalog No. 50-005-27517 Supplier MSC Supplier No. 84501543
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage is designed for precise measurement with a diameter of 0.3140 inches. It features a durable black oxide coating, providing reliable performance for various applications.
  • Precise measurement with a diameter of 0.3140 inch
  • Class ZZ Plus tolerance for accuracy
  • Durable black oxide coating for enhanced longevity