Learn More
MSC SPI Plug Gage, 0.3370 in Dia, 0.000200 in Tolerance, Black Oxide
Catalog No. 5000527345
Encompass
This plug gage is designed for precise measurement applications, featuring a black oxide finish for enhanced durability and corrosion resistance. It is manufactured from hardened drill rod for consistent performance.
  • 0.3370 inch diameter
  • 0.000200 inch tolerance
  • Black oxide coating prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length: 2 inch
Catalog No. 50-005-27345 Supplier MSC Supplier No. 84501774
Add to Cart
Add to Cart
This plug gage is designed for precise measurement applications, featuring a black oxide finish for enhanced durability and corrosion resistance. It is manufactured from hardened drill rod for consistent performance.
  • 0.3370 inch diameter
  • 0.000200 inch tolerance
  • Black oxide coating prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length: 2 inch