Learn More
MSC SPI Plug Gage, 0.3400 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000516361
Encompass
This plug gage is designed for precise measurement with a diameter of 0.3400 inch and a tolerance of 0.000200 " Constructed from steel, it provides reliable performance for various industrial applications.
  • Provides precise measurement with a 0.3400 inch diameter.
  • Offers a tight tolerance of 0.000200 inch for accuracy.
  • Made from durable steel for long-lasting use.
  • Suitable for verifying bore sizes and hole dimensions.
Catalog No. 50-005-16361 Supplier MSC Supplier No. 78385259
This plug gage is designed for precise measurement with a diameter of 0.3400 inch and a tolerance of 0.000200 " Constructed from steel, it provides reliable performance for various industrial applications.
  • Provides precise measurement with a 0.3400 inch diameter.
  • Offers a tight tolerance of 0.000200 inch for accuracy.
  • Made from durable steel for long-lasting use.
  • Suitable for verifying bore sizes and hole dimensions.