Learn More
MSC SPI Plug Gage, 0.3840 in Diameter, Class ZZ Plus, Steel
Catalog No. 5000528743
Encompass
This plug gage is designed for precise measurement, featuring a 0.3840 inch diameter. It offers high accuracy with a Class ZZ Plus tolerance of 0.000200 inch and is constructed from durable steel, making it suitable for various industrial and laboratory applications.
  • Accurate measurement of 0.3840 in diameter.
  • Class ZZ Plus tolerance for high precision.
  • Constructed from durable steel.
Catalog No. 50-005-28743 Supplier MSC Supplier No. 78385697
This plug gage is designed for precise measurement, featuring a 0.3840 inch diameter. It offers high accuracy with a Class ZZ Plus tolerance of 0.000200 inch and is constructed from durable steel, making it suitable for various industrial and laboratory applications.
  • Accurate measurement of 0.3840 in diameter.
  • Class ZZ Plus tolerance for high precision.
  • Constructed from durable steel.