Learn More
MSC SPI Plug Gage, 0.4010 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000536528
Encompass
This Class ZZ Plus Plug Gage offers precise measurement for various applications. It features a diameter of 0.4010 inches and a tight tolerance of 0.000200 inches, making it suitable for accurate inspections. The gage is constructed from durable steel.
  • Class ZZ plus plug gage
  • Diameter of measurement of 0.4010 in
  • Tolerance of 0.000200 in
  • Constructed from steel
  • Manufactured in China
Catalog No. 50-005-36528 Supplier MSC Supplier No. 78385861
This Class ZZ Plus Plug Gage offers precise measurement for various applications. It features a diameter of 0.4010 inches and a tight tolerance of 0.000200 inches, making it suitable for accurate inspections. The gage is constructed from durable steel.
  • Class ZZ plus plug gage
  • Diameter of measurement of 0.4010 in
  • Tolerance of 0.000200 in
  • Constructed from steel
  • Manufactured in China