Learn More
MSC SPI Plug Gage, 0.462 in Diameter, 0.0002 in Tolerance, Steel
Catalog No. 5000531047
Encompass
This plug gage is designed for precise measurement applications, offering high accuracy for quality control. It features a 0.462 in diameter and a 0.0002 in tolerance, crafted from durable steel for reliable performance and longevity.
  • 0.462 in diameter of measurement.
  • 0.0002 in tolerance.
  • Constructed from steel.
Catalog No. 50-005-31047 Supplier MSC Supplier No. 78386471
This plug gage is designed for precise measurement applications, offering high accuracy for quality control. It features a 0.462 in diameter and a 0.0002 in tolerance, crafted from durable steel for reliable performance and longevity.
  • 0.462 in diameter of measurement.
  • 0.0002 in tolerance.
  • Constructed from steel.