Learn More
MSC SPI Plug Gage, 0.6290 Inch, Class ZZ Minus, Steel
Catalog No. 5000530944
Encompass
This high-precision Class ZZ Minus Plug and Pin Gage is designed for accurate measurement. Manufactured from durable steel, it features a 0.6290 inch diameter with a tight tolerance of 0.0002 inch, ensuring precise inspection and quality control for various applications.
  • Constructed from steel for durability and longevity.
  • Features a precise 0.6290 inch diameter for accurate measurements.
  • Offers a tight tolerance of 0.0002 inch (Class ZZ Minus) for high-precision applications.
  • Suitable for verifying hole sizes, distances, and other critical dimensions.
Catalog No. 50-005-30944 Supplier MSC Supplier No. 78378254
This high-precision Class ZZ Minus Plug and Pin Gage is designed for accurate measurement. Manufactured from durable steel, it features a 0.6290 inch diameter with a tight tolerance of 0.0002 inch, ensuring precise inspection and quality control for various applications.
  • Constructed from steel for durability and longevity.
  • Features a precise 0.6290 inch diameter for accurate measurements.
  • Offers a tight tolerance of 0.0002 inch (Class ZZ Minus) for high-precision applications.
  • Suitable for verifying hole sizes, distances, and other critical dimensions.