Learn More
MSC SPI Plug Gage, 0.732 in, 0.0002 in Tolerance, Class ZZ Plus
Catalog No. 5000530135
Encompass
This plug gage is designed for precise measurement applications. It features a 0.732 inch diameter and a tight 0.0002 inch tolerance, making it suitable for verifying hole dimensions with high accuracy. Constructed from durable steel, this Class ZZ Plus gage ensures reliable performance.
  • 0.732 inch diameter for accurate measurements.
  • 0.0002 inch tolerance for high precision.
  • Constructed from steel for durability.
  • Class ZZ Plus for quality assurance.
Catalog No. 50-005-30135 Supplier MSC Supplier No. 78389178
Add to Cart
Add to Cart
This plug gage is designed for precise measurement applications. It features a 0.732 inch diameter and a tight 0.0002 inch tolerance, making it suitable for verifying hole dimensions with high accuracy. Constructed from durable steel, this Class ZZ Plus gage ensures reliable performance.
  • 0.732 inch diameter for accurate measurements.
  • 0.0002 inch tolerance for high precision.
  • Constructed from steel for durability.
  • Class ZZ Plus for quality assurance.