Learn More
MSC SPI Plug Gage, Class ZZ, 0.3630 in Diameter, 0.000200 in Tolerance
Catalog No. 5000528752
Encompass
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate verification of hole diameters. Made from durable steel, it features a precise diameter of 0.3630 inches and a tight tolerance of 0.000200 inches, ensuring reliable measurements for quality control and manufacturing applications.
  • Class ZZ plus plug gage for precise measurement
  • Diameter of 0.3630 inch
  • Tolerance of 0.000200 inch
  • Constructed from high-quality steel for durability
Catalog No. 50-005-28752 Supplier MSC Supplier No. 78385481
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate verification of hole diameters. Made from durable steel, it features a precise diameter of 0.3630 inches and a tight tolerance of 0.000200 inches, ensuring reliable measurements for quality control and manufacturing applications.
  • Class ZZ plus plug gage for precise measurement
  • Diameter of 0.3630 inch
  • Tolerance of 0.000200 inch
  • Constructed from high-quality steel for durability