Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0310 in Dia, Black Oxide
Catalog No. 5000515670
Encompass
This plug gage is designed for precise measurement applications, featuring a Class ZZ Plus tolerance for accuracy. It has a 0.0310 in diameter and a durable black oxide finish, making it suitable for checking bore sizes and hole locations.
  • Class ZZ Plus tolerance for high accuracy
  • 0.0310 in diameter for precise measurements
  • Black oxide finish provides durability
  • 0.000200 in tolerance ensures reliable performance
Catalog No. 50-005-15670 Supplier MSC Supplier No. 84498708
This plug gage is designed for precise measurement applications, featuring a Class ZZ Plus tolerance for accuracy. It has a 0.0310 in diameter and a durable black oxide finish, making it suitable for checking bore sizes and hole locations.
  • Class ZZ Plus tolerance for high accuracy
  • 0.0310 in diameter for precise measurements
  • Black oxide finish provides durability
  • 0.000200 in tolerance ensures reliable performance