Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0310 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000516424
Encompass
This plug gage is a Class ZZ Plus tool designed for precise measurements. It features a specific diameter and high tolerance, making it suitable for demanding quality control and inspection tasks. Constructed from durable steel, it ensures reliability and longevity in various industrial and scientific applications.
  • Class ZZ Plus for high-precision applications
  • 0.0310 inch diameter for accurate sizing
  • 0.000200 inch tolerance ensures high accuracy
  • Made from steel for durability
Catalog No. 50-005-16424 Supplier MSC Supplier No. 78382173
This plug gage is a Class ZZ Plus tool designed for precise measurements. It features a specific diameter and high tolerance, making it suitable for demanding quality control and inspection tasks. Constructed from durable steel, it ensures reliability and longevity in various industrial and scientific applications.
  • Class ZZ Plus for high-precision applications
  • 0.0310 inch diameter for accurate sizing
  • 0.000200 inch tolerance ensures high accuracy
  • Made from steel for durability