Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0350 inch Diameter, Steel
Catalog No. 5000528824
Encompass
This plug gage is designed for precise measurement applications, featuring a 0.0350 inch diameter and a tight 0.000200 inch tolerance. Manufactured from durable steel, it is suitable for verifying hole sizes and ensuring dimensional accuracy in various industrial and quality control settings.
  • Diameter of 0.0350 inch
  • Tolerance of 0.000200 inch for high precision
  • Constructed from steel for durability
  • Suitable for accurate hole size verification
Catalog No. 50-005-28824 Supplier MSC Supplier No. 78382215
This plug gage is designed for precise measurement applications, featuring a 0.0350 inch diameter and a tight 0.000200 inch tolerance. Manufactured from durable steel, it is suitable for verifying hole sizes and ensuring dimensional accuracy in various industrial and quality control settings.
  • Diameter of 0.0350 inch
  • Tolerance of 0.000200 inch for high precision
  • Constructed from steel for durability
  • Suitable for accurate hole size verification