Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0480 in Dia, Steel
Catalog No. 5000527264
Encompass
This plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, ensuring accuracy for quality control and inspection tasks. Constructed from durable steel, it offers reliability in various industrial settings.
  • Features a class ZZ Plus tolerance.
  • Has a measurement diameter of 0.0480 inches.
  • Provides a tolerance of 0.000200 inches.
  • Made from steel for durability.
  • Manufactured in China.
Catalog No. 50-005-27264 Supplier MSC Supplier No. 78382348
Add to Cart
Add to Cart
This plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, ensuring accuracy for quality control and inspection tasks. Constructed from durable steel, it offers reliability in various industrial settings.
  • Features a class ZZ Plus tolerance.
  • Has a measurement diameter of 0.0480 inches.
  • Provides a tolerance of 0.000200 inches.
  • Made from steel for durability.
  • Manufactured in China.