Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0580 in Diameter, 0.0002 in Tolerance, Steel
Catalog No. 5000527265
Encompass
The SPI Class ZZ Plus Plug Gage is a precision measuring tool designed for accurately checking the diameter of holes. Made from durable steel, this gage features a 0.0580 inch diameter and a tight 0.0002 inch tolerance, ensuring high accuracy for quality control and inspection applications.
  • Class ZZ Plus
  • 0.0580 in diameter measurement
  • 0.0002 in tolerance
  • Constructed from steel
Catalog No. 50-005-27265 Supplier MSC Supplier No. 78382439
The SPI Class ZZ Plus Plug Gage is a precision measuring tool designed for accurately checking the diameter of holes. Made from durable steel, this gage features a 0.0580 inch diameter and a tight 0.0002 inch tolerance, ensuring high accuracy for quality control and inspection applications.
  • Class ZZ Plus
  • 0.0580 in diameter measurement
  • 0.0002 in tolerance
  • Constructed from steel