Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0910 in Diameter, Black Oxide
Catalog No. 5000527594
Encompass
This Class ZZ Plus plug gage features a 0.0910 inch diameter and a 0.000200 inch tolerance. It is made from durable tool steel and finished with a black oxide coating.
  • For go inspection.
  • Can be used to set micrometers.
  • Can be used to set verniers.
  • Can be used to set other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-27594 Supplier MSC Supplier No. 84499300
Add to Cart
Add to Cart
This Class ZZ Plus plug gage features a 0.0910 inch diameter and a 0.000200 inch tolerance. It is made from durable tool steel and finished with a black oxide coating.
  • For go inspection.
  • Can be used to set micrometers.
  • Can be used to set verniers.
  • Can be used to set other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.