Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0960 in Dia, 0.0002 in Tolerance
Catalog No. 5000531506
Encompass
This plug gage is a Class ZZ Plus precision measuring tool. It features a 0.0960 inch diameter and a tight 0.000200 inch tolerance, with a black oxide coating for enhanced durability and corrosion resistance.
  • Class ZZ Plus precision for accurate measurements.
  • 0.0960 inch diameter for specific sizing applications.
  • 0.000200 inch tolerance ensures high accuracy.
  • Black oxide coating provides durability and corrosion resistance.
  • Ideal for calibration and quality control tasks.
Catalog No. 50-005-31506 Supplier MSC Supplier No. 84499359
This plug gage is a Class ZZ Plus precision measuring tool. It features a 0.0960 inch diameter and a tight 0.000200 inch tolerance, with a black oxide coating for enhanced durability and corrosion resistance.
  • Class ZZ Plus precision for accurate measurements.
  • 0.0960 inch diameter for specific sizing applications.
  • 0.000200 inch tolerance ensures high accuracy.
  • Black oxide coating provides durability and corrosion resistance.
  • Ideal for calibration and quality control tasks.