Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.0970 inch Diameter, Steel
Catalog No. 5000530837
Encompass
This plug gage is a Class ZZ Plus instrument featuring a 0.0970 in diameter and a 0.000200 in tolerance. Constructed from steel, it is suitable for precise measurement applications.
  • Class ZZ Plus for high precision
  • 0.0970 inch measurement diameter
  • Tight tolerance of 0.000200 inch
  • Durable steel construction
Catalog No. 50-005-30837 Supplier MSC Supplier No. 78382827
This plug gage is a Class ZZ Plus instrument featuring a 0.0970 in diameter and a 0.000200 in tolerance. Constructed from steel, it is suitable for precise measurement applications.
  • Class ZZ Plus for high precision
  • 0.0970 inch measurement diameter
  • Tight tolerance of 0.000200 inch
  • Durable steel construction