Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.1190 in Diameter, Black Oxide
Catalog No. 5000515258
Encompass
This plug gage is designed for precise measurement applications. It features a Class ZZ Plus tolerance, a 0.1190 in diameter, and a durable black oxide finish, with a tolerance of 0.000200 in.
  • Features Class ZZ Plus
  • Designed for 0.1190 in diameter measurements
  • Durable black oxide finish
  • Ensures 0.000200 in tolerance
Catalog No. 50-005-15258 Supplier MSC Supplier No. 84499581
This plug gage is designed for precise measurement applications. It features a Class ZZ Plus tolerance, a 0.1190 in diameter, and a durable black oxide finish, with a tolerance of 0.000200 in.
  • Features Class ZZ Plus
  • Designed for 0.1190 in diameter measurements
  • Durable black oxide finish
  • Ensures 0.000200 in tolerance