Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.1490 in Diameter, 0.000200 in Tolerance
Supplier:  MSC 78383346
Encompass
This plug gage is designed for precise measurements, featuring a 0.1490 in diameter and a 0.000200 in tolerance. Constructed from durable steel, it is suitable for various industrial applications requiring accurate dimension verification.
  • Class ZZ plus plug gage
  • 0.1490 inch diameter
  • 0.000200 inch tolerance
  • Made of steel
  • Country of origin is China
Catalog No. 50-005-30792