Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.1800 in Dia, Steel
Catalog No. 5000530529
Encompass
This plug gage is a Class ZZ Plus precision tool designed for accurate measurement. It features a 0.1800 inch diameter and a tolerance of 0.000200 inch, making it suitable for demanding applications requiring high precision. It is constructed from durable steel.
  • Class ZZ Plus for precision measurement
  • 0.1800 inch diameter for specific sizing needs
  • Tolerance of 0.000200 inch ensures high accuracy
  • Made from steel for durability
Catalog No. 50-005-30529 Supplier MSC Supplier No. 78383650
This plug gage is a Class ZZ Plus precision tool designed for accurate measurement. It features a 0.1800 inch diameter and a tolerance of 0.000200 inch, making it suitable for demanding applications requiring high precision. It is constructed from durable steel.
  • Class ZZ Plus for precision measurement
  • 0.1800 inch diameter for specific sizing needs
  • Tolerance of 0.000200 inch ensures high accuracy
  • Made from steel for durability