Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.1940 Inch Diameter, Steel
Catalog No. 5000516360
Encompass
SPI Class ZZ Plus Plug and Pin Gages are precision measuring tools designed for accurate diameter verification. Made from durable steel, these gages offer reliable performance for various industrial applications.
  • Diameter of 0.1940 inch
  • Tolerance of 0.000200 decimal inch (0.0002 mm)
  • Constructed from steel for durability
  • Includes a factory warranty
Catalog No. 50-005-16360 Supplier MSC Supplier No. 78383791
SPI Class ZZ Plus Plug and Pin Gages are precision measuring tools designed for accurate diameter verification. Made from durable steel, these gages offer reliable performance for various industrial applications.
  • Diameter of 0.1940 inch
  • Tolerance of 0.000200 decimal inch (0.0002 mm)
  • Constructed from steel for durability
  • Includes a factory warranty