Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2040 in Diameter, Black Oxide Finish
Catalog No. 5000527595
Encompass
This plug gage is a precision measuring tool, featuring a 0.2040 inch diameter and a durable black oxide finish. It is designed for accurate measurement with a tight tolerance of 0.000200 inch, making it suitable for quality control and inspection applications.
  • Features a 0.2040 inch diameter.
  • Has a black oxide finish for enhanced durability.
  • Offers a precise tolerance of 0.000200 inch for accurate measurements.
  • Manufactured in China.
  • Includes a hassle-free guarantee for exchanges or refunds within 30 days of shipment (exclusions apply).
Catalog No. 50-005-27595 Supplier MSC Supplier No. 84500438
This plug gage is a precision measuring tool, featuring a 0.2040 inch diameter and a durable black oxide finish. It is designed for accurate measurement with a tight tolerance of 0.000200 inch, making it suitable for quality control and inspection applications.
  • Features a 0.2040 inch diameter.
  • Has a black oxide finish for enhanced durability.
  • Offers a precise tolerance of 0.000200 inch for accurate measurements.
  • Manufactured in China.
  • Includes a hassle-free guarantee for exchanges or refunds within 30 days of shipment (exclusions apply).