Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2080 in Diameter, Steel
Catalog No. 5000528831
Encompass
The SPI Class ZZ Plus Plug Gage provides precise dimensional verification for industrial and scientific applications. Constructed from durable steel, this gage features a diameter of 0.2080 inch and a tight tolerance of 0.0002 "
  • Provides precise dimensional verification.
  • Constructed from durable steel.
  • Features a tight tolerance for accuracy.
  • Suitable for various industrial and scientific applications.
Catalog No. 50-005-28831 Supplier MSC Supplier No. 78383932
The SPI Class ZZ Plus Plug Gage provides precise dimensional verification for industrial and scientific applications. Constructed from durable steel, this gage features a diameter of 0.2080 inch and a tight tolerance of 0.0002 "
  • Provides precise dimensional verification.
  • Constructed from durable steel.
  • Features a tight tolerance for accuracy.
  • Suitable for various industrial and scientific applications.