Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2100 inch Diameter, Steel
Catalog No. 5000511645
Encompass
This plug gage is engineered for precise measurement applications. It features a Class ZZ Plus tolerance and a 0.2100 inch diameter, making it suitable for accurate verification of hole sizes.
  • Class ZZ Plus tolerance for high accuracy.
  • 0.2100 inch diameter for specific measurement needs.
  • Made from durable steel for long-lasting performance.
  • Ideal for quality control and inspection tasks.
  • Ensures accurate hole sizing.
Catalog No. 50-005-11645 Supplier MSC Supplier No. 78383957
This plug gage is engineered for precise measurement applications. It features a Class ZZ Plus tolerance and a 0.2100 inch diameter, making it suitable for accurate verification of hole sizes.
  • Class ZZ Plus tolerance for high accuracy.
  • 0.2100 inch diameter for specific measurement needs.
  • Made from durable steel for long-lasting performance.
  • Ideal for quality control and inspection tasks.
  • Ensures accurate hole sizing.