Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2670 in Dia, Black Oxide
Catalog No. 5000527600
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.2670 inch diameter and a tight tolerance of 0.000200 inch, making it suitable for verifying critical dimensions. The gage is finished with a black oxide coating for enhanced durability.
  • Class ZZ Plus gage for accurate go/no-go measurement.
  • 0.2670 inch diameter ensures precise fit.
  • Tolerance of 0.000200 inch for high-precision inspection.
  • Black oxide coating provides durability and corrosion resistance.
Catalog No. 50-005-27600 Supplier MSC Supplier No. 84501063
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.2670 inch diameter and a tight tolerance of 0.000200 inch, making it suitable for verifying critical dimensions. The gage is finished with a black oxide coating for enhanced durability.
  • Class ZZ Plus gage for accurate go/no-go measurement.
  • 0.2670 inch diameter ensures precise fit.
  • Tolerance of 0.000200 inch for high-precision inspection.
  • Black oxide coating provides durability and corrosion resistance.