Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2670 in, Steel
Catalog No. 5000530825
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.2670 inch diameter and is constructed from steel for durability and consistent performance in various applications.
  • Class ZZ Plus
  • Diameter of 0.2670 inch
  • Tolerance of 0.000200 inch
  • Made from steel
Catalog No. 50-005-30825 Supplier MSC Supplier No. 78384526
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.2670 inch diameter and is constructed from steel for durability and consistent performance in various applications.
  • Class ZZ Plus
  • Diameter of 0.2670 inch
  • Tolerance of 0.000200 inch
  • Made from steel