Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.2800 in Diameter, 0.0002 in Tolerance
Catalog No. 5000528836
Encompass
This is a Class ZZ Plus plug gage designed for precise measurement. It is engineered to provide accurate and reliable results for various inspection applications.
  • Class ZZ Plus grade for high precision
  • Features a 0.2800 in diameter
  • Manufactured with a 0.0002 in tolerance
Catalog No. 50-005-28836 Supplier MSC Supplier No. 78384658
This is a Class ZZ Plus plug gage designed for precise measurement. It is engineered to provide accurate and reliable results for various inspection applications.
  • Class ZZ Plus grade for high precision
  • Features a 0.2800 in diameter
  • Manufactured with a 0.0002 in tolerance