Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.3150 in Dia, Black Oxide
Catalog No. 5000531738
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3150 inch diameter and a tight tolerance of 0.000200 inches, with a durable black oxide finish.
  • Class ZZ Plus gage
  • 0.3150 inch diameter
  • 0.000200 inch tolerance
  • Black oxide finish
  • Country of origin is China
Catalog No. 50-005-31738 Supplier MSC Supplier No. 62255195
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3150 inch diameter and a tight tolerance of 0.000200 inches, with a durable black oxide finish.
  • Class ZZ Plus gage
  • 0.3150 inch diameter
  • 0.000200 inch tolerance
  • Black oxide finish
  • Country of origin is China