Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.3960 inch Diameter, Black Oxide Finish
Catalog No. 5000531416
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3960 inch diameter, a 0.000200 inch tolerance, and a black oxide finish for durability.
  • Designed for precise measurement applications
  • Features a 0.3960 inch diameter
  • Provides a 0.000200 inch tolerance
  • Durable black oxide finish
  • Manufactured in China
Catalog No. 50-005-31416 Supplier MSC Supplier No. 84502368
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.3960 inch diameter, a 0.000200 inch tolerance, and a black oxide finish for durability.
  • Designed for precise measurement applications
  • Features a 0.3960 inch diameter
  • Provides a 0.000200 inch tolerance
  • Durable black oxide finish
  • Manufactured in China