Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.3990 in, Steel
Catalog No. 5000528828
Encompass
This plug gage is designed for precise measurement and inspection. It is a Class ZZ Plus gage with a diameter of 0.3990 inches and a tolerance of 0.000200 inches, ensuring high accuracy. Constructed from durable steel, this gage is suitable for various industrial applications requiring tight dimensional control.
  • Class ZZ Plus designation
  • Features a 0.3990 inch diameter
  • Provides a tolerance of 0.000200 inch
  • Made from steel for durability
  • Ideal for accurate dimensional verification
Catalog No. 50-005-28828 Supplier MSC Supplier No. 78385846
This plug gage is designed for precise measurement and inspection. It is a Class ZZ Plus gage with a diameter of 0.3990 inches and a tolerance of 0.000200 inches, ensuring high accuracy. Constructed from durable steel, this gage is suitable for various industrial applications requiring tight dimensional control.
  • Class ZZ Plus designation
  • Features a 0.3990 inch diameter
  • Provides a tolerance of 0.000200 inch
  • Made from steel for durability
  • Ideal for accurate dimensional verification