Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.4430 in, 0.0002 in Tolerance
Catalog No. 5000531060
Encompass
This plug and pin gage is a Class ZZ Plus tool designed for precise measurements. It features a 0.4430 inch diameter and a tolerance of 0.0002 " Constructed from steel, this made-to-order item is suitable for various applications requiring high-precision gaging. Please note that all sales are final, and the product is not eligible for return or exchange unless defective, with an average lead time of 1 to 2 weeks. A minimum order of $100 is required.
  • Diameter of 0.4430 inch
  • Tolerance of 0.0002 inch
  • Constructed from steel
  • Class ZZ Plus designation for precision
  • Made-to-order item
  • Not eligible for return or exchange unless defective
  • Average lead time of 1 to 2 weeks
  • Minimum order requirement of $100
Catalog No. 50-005-31060 Supplier MSC Supplier No. 78386281
Add to Cart
Add to Cart
This plug and pin gage is a Class ZZ Plus tool designed for precise measurements. It features a 0.4430 inch diameter and a tolerance of 0.0002 " Constructed from steel, this made-to-order item is suitable for various applications requiring high-precision gaging. Please note that all sales are final, and the product is not eligible for return or exchange unless defective, with an average lead time of 1 to 2 weeks. A minimum order of $100 is required.
  • Diameter of 0.4430 inch
  • Tolerance of 0.0002 inch
  • Constructed from steel
  • Class ZZ Plus designation for precision
  • Made-to-order item
  • Not eligible for return or exchange unless defective
  • Average lead time of 1 to 2 weeks
  • Minimum order requirement of $100