Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.455 in Diameter, Steel
Catalog No. 5000529013
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.455 in diameter and a tight 0.0002 in tolerance, constructed from durable steel.
  • Designed for precise measurement applications
  • Features a 0.455 in diameter
  • Provides a 0.0002 in tolerance
  • Constructed from durable steel
Catalog No. 50-005-29013 Supplier MSC Supplier No. 78386406
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.455 in diameter and a tight 0.0002 in tolerance, constructed from durable steel.
  • Designed for precise measurement applications
  • Features a 0.455 in diameter
  • Provides a 0.0002 in tolerance
  • Constructed from durable steel