Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.4640 inch Diameter, Steel
Catalog No. 5000530895
Encompass
This plug gage is designed for precise measurement applications. It features a specific diameter and a tight tolerance, making it suitable for quality control and calibration tasks in various industries.
  • Class ZZ Plus tolerance
  • 0.4640 inch diameter of measurement
  • 0.000200 inch tolerance
  • Constructed from steel
Catalog No. 50-005-30895 Supplier MSC Supplier No. 78386497
This plug gage is designed for precise measurement applications. It features a specific diameter and a tight tolerance, making it suitable for quality control and calibration tasks in various industries.
  • Class ZZ Plus tolerance
  • 0.4640 inch diameter of measurement
  • 0.000200 inch tolerance
  • Constructed from steel