Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.474 in Diameter, 0.0002 in Tolerance
Catalog No. 5000530929
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.474 in diameter and a tolerance of 0.0002 in, ensuring high accuracy for inspecting bore sizes. Constructed from durable steel, this gage provides reliability and longevity in various industrial environments.
  • High precision for accurate measurements.
  • Durable steel construction for extended lifespan.
  • Suitable for inspecting bore sizes.
  • Reliable performance in industrial settings.
Catalog No. 50-005-30929 Supplier MSC Supplier No. 78386596
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.474 in diameter and a tolerance of 0.0002 in, ensuring high accuracy for inspecting bore sizes. Constructed from durable steel, this gage provides reliability and longevity in various industrial environments.
  • High precision for accurate measurements.
  • Durable steel construction for extended lifespan.
  • Suitable for inspecting bore sizes.
  • Reliable performance in industrial settings.