Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.4900 in Diameter, 0.000200 in Tolerance
Catalog No. 5000527268
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications, featuring a 0.4900 inch diameter and a tight 0.000200 inch tolerance. Constructed from steel, it is suitable for various industrial and laboratory calibration tasks.
  • Class ZZ Plus plug gage for precise measurement
  • 0.4900 inch diameter for accurate gauging
  • 0.000200 inch tolerance ensures high precision
  • Durable steel construction for longevity
Catalog No. 50-005-27268 Supplier MSC Supplier No. 78386752
This Class ZZ Plus Plug Gage is designed for precise measurement applications, featuring a 0.4900 inch diameter and a tight 0.000200 inch tolerance. Constructed from steel, it is suitable for various industrial and laboratory calibration tasks.
  • Class ZZ Plus plug gage for precise measurement
  • 0.4900 inch diameter for accurate gauging
  • 0.000200 inch tolerance ensures high precision
  • Durable steel construction for longevity