Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.4920 in Diameter, 0.0002 in Tolerance
Catalog No. 5000529004
Encompass
This plug gage is a Class ZZ Plus model, designed for precise measurement applications. It features a 0.4920 inch diameter and a tight 0.0002 inch tolerance, ensuring accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory environments.
  • Class ZZ Plus designation
  • 0.4920 inch diameter
  • 0.0002 inch tolerance
  • Durable steel construction
Catalog No. 50-005-29004 Supplier MSC Supplier No. 78386778
Add to Cart
Add to Cart
This plug gage is a Class ZZ Plus model, designed for precise measurement applications. It features a 0.4920 inch diameter and a tight 0.0002 inch tolerance, ensuring accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory environments.
  • Class ZZ Plus designation
  • 0.4920 inch diameter
  • 0.0002 inch tolerance
  • Durable steel construction