Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.5050 in Diameter, Black Oxide
Catalog No. 5000527603
Encompass
This plug gage is a Class ZZ Plus tool with a 0.5050 inch diameter and a 0.000200 inch tolerance, featuring a black oxide finish. It is suitable for Go Inspection and calibrating measuring devices. A Certificate of Accuracy with NIST Traceability is included.
  • Class ZZ Plus
  • 0.5050 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Includes Certificate of Accuracy with NIST Traceability
  • Used for Go Inspection
  • Can be used to set micrometers, verniers, and other measuring devices
Catalog No. 50-005-27603 Supplier MSC Supplier No. 84503457
Add to Cart
Add to Cart
This plug gage is a Class ZZ Plus tool with a 0.5050 inch diameter and a 0.000200 inch tolerance, featuring a black oxide finish. It is suitable for Go Inspection and calibrating measuring devices. A Certificate of Accuracy with NIST Traceability is included.
  • Class ZZ Plus
  • 0.5050 in diameter
  • 0.000200 in tolerance
  • Black oxide finish
  • Includes Certificate of Accuracy with NIST Traceability
  • Used for Go Inspection
  • Can be used to set micrometers, verniers, and other measuring devices