Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.5520 in Diameter, 0.000200 in Tolerance
Catalog No. 5000528832
Encompass
This precision plug gage accurately checks hole diameters. With a 0.5520 in diameter and 0.000200 in tolerance, it ensures precise measurements for quality control in manufacturing and machining. Made from durable steel, it provides reliability and longevity. Ideal for verifying hole sizes and ensuring specifications, its "Plus" classification suits maximum material conditions or GO gauging.
  • Accurately checks hole diameters.
  • Features a 0.5520 in diameter and 0.000200 in tolerance.
  • Ensures precise measurements for quality control.
  • Durable steel construction provides reliability and longevity.
  • Verifies hole sizes and meets exact specifications.
  • "Plus" classification for maximum material conditions or GO gauging.
Catalog No. 50-005-28832 Supplier MSC Supplier No. 78387370
This precision plug gage accurately checks hole diameters. With a 0.5520 in diameter and 0.000200 in tolerance, it ensures precise measurements for quality control in manufacturing and machining. Made from durable steel, it provides reliability and longevity. Ideal for verifying hole sizes and ensuring specifications, its "Plus" classification suits maximum material conditions or GO gauging.
  • Accurately checks hole diameters.
  • Features a 0.5520 in diameter and 0.000200 in tolerance.
  • Ensures precise measurements for quality control.
  • Durable steel construction provides reliability and longevity.
  • Verifies hole sizes and meets exact specifications.
  • "Plus" classification for maximum material conditions or GO gauging.