Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.5520 inch Diameter, Black Oxide
Catalog No. 5000530434
Encompass
This plug gage is designed for precise measurement applications, featuring a 0.5520 inch diameter for accurate hole sizing. It comes with a Class ZZ Plus tolerance of +0.000200 inch, ensuring high precision. The black oxide finish provides durability and corrosion resistance.
  • Designed for precise measurement applications
  • Accurate 0.5520 inch diameter
  • Class ZZ Plus tolerance of +0.000200 inch
  • Black oxide finish for durability and corrosion resistance
Catalog No. 50-005-30434 Supplier MSC Supplier No. 56239031
This plug gage is designed for precise measurement applications, featuring a 0.5520 inch diameter for accurate hole sizing. It comes with a Class ZZ Plus tolerance of +0.000200 inch, ensuring high precision. The black oxide finish provides durability and corrosion resistance.
  • Designed for precise measurement applications
  • Accurate 0.5520 inch diameter
  • Class ZZ Plus tolerance of +0.000200 inch
  • Black oxide finish for durability and corrosion resistance