Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.603 Inch Diameter, Black Oxide Finish
Catalog No. 5000527662
Encompass
This plug gage is designed for precise measurement and quality control, ensuring accuracy in various applications. It features a Class ZZ Plus tolerance, a 0.603 inch diameter, and a durable black oxide finish for extended tool life and resistance to corrosion.
  • Class ZZ Plus tolerance for high precision measurements.
  • 0.603 inch diameter for accurate gauging.
  • Black oxide finish for enhanced durability and corrosion resistance.
  • Tolerance of +0.000200 inch for critical dimensions.
  • Tolerance of 0.0002 mm for metric equivalent precision.
Catalog No. 50-005-27662 Supplier MSC Supplier No. 56239551
This plug gage is designed for precise measurement and quality control, ensuring accuracy in various applications. It features a Class ZZ Plus tolerance, a 0.603 inch diameter, and a durable black oxide finish for extended tool life and resistance to corrosion.
  • Class ZZ Plus tolerance for high precision measurements.
  • 0.603 inch diameter for accurate gauging.
  • Black oxide finish for enhanced durability and corrosion resistance.
  • Tolerance of +0.000200 inch for critical dimensions.
  • Tolerance of 0.0002 mm for metric equivalent precision.