Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6130 inch Diameter, Black Oxide
Catalog No. 5000531281
Encompass
This product is a Class ZZ Plus Plug Gage, featuring a 0.6130 inch diameter and a durable black oxide finish. It is designed for precise measurement applications, offering reliable performance for quality control and calibration tasks.
  • Features a tolerance of +0.000200 "
  • Coating with black oxide for enhanced durability and corrosion resistance.
  • Precisely manufactured with a 0.6130 inch diameter.
Catalog No. 50-005-31281 Supplier MSC Supplier No. 56239650
This product is a Class ZZ Plus Plug Gage, featuring a 0.6130 inch diameter and a durable black oxide finish. It is designed for precise measurement applications, offering reliable performance for quality control and calibration tasks.
  • Features a tolerance of +0.000200 "
  • Coating with black oxide for enhanced durability and corrosion resistance.
  • Precisely manufactured with a 0.6130 inch diameter.