Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6220 in Diameter, Black Oxide Finish
Catalog No. 5000530829
Encompass
This plug gage is a precision measuring tool designed for accurate verification of bore diameters. It features a 0.6220 inch diameter with a Class ZZ Plus tolerance, ensuring high accuracy for critical applications. The black oxide finish provides enhanced durability.
  • Class ZZ Plus tolerance, with a +0.000200 in tolerance.
  • 0.6220 inch diameter.
  • Black oxide finish for durability.
  • Suitable for checking bore diameters.
Catalog No. 50-005-30829 Supplier MSC Supplier No. 78388071
This plug gage is a precision measuring tool designed for accurate verification of bore diameters. It features a 0.6220 inch diameter with a Class ZZ Plus tolerance, ensuring high accuracy for critical applications. The black oxide finish provides enhanced durability.
  • Class ZZ Plus tolerance, with a +0.000200 in tolerance.
  • 0.6220 inch diameter.
  • Black oxide finish for durability.
  • Suitable for checking bore diameters.