Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6320" Dia, Black Oxide
Catalog No. 5000514711
Encompass
This plug gage is a Class ZZ Plus with a 0.6320" diameter. It features a +0.000200" tolerance and a black oxide finish, making it suitable for precise measurement applications.
  • Class ZZ Plus Plug Gage
  • 0.6320" diameter
  • +0.000200" tolerance
  • Black oxide finish
Catalog No. 50-005-14711 Supplier MSC Supplier No. 56239841
This plug gage is a Class ZZ Plus with a 0.6320" diameter. It features a +0.000200" tolerance and a black oxide finish, making it suitable for precise measurement applications.
  • Class ZZ Plus Plug Gage
  • 0.6320" diameter
  • +0.000200" tolerance
  • Black oxide finish