Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6340 in Diameter, Steel
Catalog No. 5000516365
Encompass
This Class ZZ Plus plug gage offers precise measurements with a 0.6340 in diameter and a tolerance of 0.000200 in, ideal for accurate inspection and quality control.
  • Class ZZ Plus
  • Diameter of measurement: 0.6340 in
  • Tolerance: 0.000200 in
  • Material: Steel
Catalog No. 50-005-16365 Supplier MSC Supplier No. 78388196
This Class ZZ Plus plug gage offers precise measurements with a 0.6340 in diameter and a tolerance of 0.000200 in, ideal for accurate inspection and quality control.
  • Class ZZ Plus
  • Diameter of measurement: 0.6340 in
  • Tolerance: 0.000200 in
  • Material: Steel